Assay Details
| Gene Symbol: dscama | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030224.2 | NP_001025395.1 | 32 - 33 | 6276 | 80 | |
| XM_005155472.3 | XP_005155529.1 | 26 - 27 | 4350 | 80 | ||
| XM_021479191.1 | XP_021334866.1 | 32 - 33 | 6353 | 80 | ||
| GenBank mRNA | AY552547.1 | - | 32 - 33 | 6276 | 80 | |
More Information
Set Membership: |
|

dscama
Genomic Map