Assay Details
| Gene Symbol: hmbox1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030264.1 | NP_001025435.1 | 3 - 4 | 824 | 74 | |
| XM_005169831.4 | XP_005169888.1 | 3 - 4 | 739 | 74 | ||
| XM_005169832.4 | XP_005169889.1 | 3 - 4 | 932 | 74 | ||
| XM_009292894.3 | XP_009291169.1 | 3 - 4 | 937 | 74 | ||
| XM_021467220.1 | XP_021322895.1 | 3 - 4 | 936 | 74 | ||
| XM_021467221.1 | XP_021322896.1 | 3 - 4 | 933 | 74 | ||
| GenBank mRNA | BC097126.1 | - | 3 - 4 | 824 | 74 | |
More Information
Set Membership: |
|

hmbox1a
Genomic Map