Assay Details
| Gene Symbol: hmbox1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030264.1 | NP_001025435.1 | 7 - 8 | 1282 | 74 | |
| XM_021467221.1 | XP_021322896.1 | 7 - 8 | 1391 | 74 | ||
| GenBank mRNA | BC097126.1 | - | 7 - 8 | 1282 | 74 | |
More Information
Set Membership: |
|

hmbox1a
Genomic Map