Assay Details
| Gene Symbol: hmbox1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030264.1 | NP_001025435.1 | 8 - 9 | 1390 | 99 | |
| XM_005169831.4 | XP_005169888.1 | 8 - 9 | 1311 | 99 | ||
| XM_005169832.4 | XP_005169889.1 | 8 - 9 | 1501 | 99 | ||
| GenBank mRNA | BC097126.1 | - | 8 - 9 | 1390 | 99 | |
More Information
Set Membership: |
|

hmbox1a
Genomic Map