Assay Details
| Gene Symbol: itgb1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034987.1 | NP_001030159.1 | 1917 | 70 | ||
| GenBank mRNA | BC162706.1 | - | 2014 | 70 | ||
| DQ149102.1 | - | 1917 | 70 | |||
| HQ538840.1 | - | 2187 | 70 | |||
More Information
Set Membership: |
|

itgb1b
Genomic Map