Assay Details
| Gene Symbol: itgb1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034987.1 | NP_001030159.1 | 141 | 70 | ||
| GenBank mRNA | BC162706.1 | - | 238 | 70 | ||
| DQ149102.1 | - | 141 | 70 | |||
| HQ538840.1 | - | 411 | 70 | |||
| Gene Symbol: LOC110437890 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021466326.1 | XP_021322001.1 | 3 - 4 | 481 | 70 | |
More Information
Set Membership: |
|

itgb1b
Genomic Map