Assay Details
| Gene Symbol: apooa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037115.1 | NP_001032192.1 | 6 - 7 | 523 | 56 | |
| GenBank mRNA | BC107823.1 | - | 6 - 7 | 523 | 56 | |
More Information
Set Membership: |
|

apooa
Genomic Map