Assay Details
| Gene Symbol: naga | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037388.2 | NP_001032465.2 | 6 - 7 | 904 | 64 | |
| XM_021475044.1 | XP_021330719.1 | 6 - 7 | 851 | 64 | ||
| GenBank mRNA | BC108078.1 | - | 6 - 7 | 872 | 64 | |
| BC139643.1 | - | 6 - 7 | 819 | 64 | ||
| EH434772.1 | - | 6 - 7 | 807 | 64 | ||
More Information
Set Membership: |
|

naga
Genomic Map