Assay Details
| Gene Symbol: ppp2r2ca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037394.1 | NP_001032471.1 | 6 - 7 | 851 | 66 | |
| XM_021478641.1 | XP_021334316.1 | 5 - 6 | 910 | 66 | ||
| GenBank mRNA | BC109406.1 | - | 6 - 7 | 851 | 66 | |
More Information
Set Membership: |
|

ppp2r2ca
Genomic Map