Assay Details
| Gene Symbol: itgb1bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037409.1 | NP_001032486.1 | 2 - 3 | 253 | 60 | |
| GenBank mRNA | BC108035.1 | - | 2 - 3 | 253 | 60 | |
More Information
Set Membership: |
|

itgb1bp1
Genomic Map