Assay Details
| Gene Symbol: reep3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037575.1 | NP_001032664.1 | 3 - 4 | 430 | 102 | |
| GenBank mRNA | BC110095.1 | - | 3 - 4 | 430 | 102 | |
More Information
Set Membership: |
|

reep3a
Genomic Map