Assay Details
| Gene Symbol: pstpip1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037579.1 | NP_001032668.1 | 10 - 11 | 842 | 82 | |
| GenBank mRNA | BC110107.1 | - | 10 - 11 | 842 | 82 | |
More Information
Set Membership: |
|

pstpip1b
Genomic Map