Assay Details
| Gene Symbol: epoa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038009.2 | NP_001033098.1 | 2 - 3 | 441 | 78 | |
| NM_001115127.1 | NP_001108599.1 | 3 - 4 | 352 | 78 | ||
| NM_001115128.1 | NP_001108600.1 | 2 - 3 | 173 | 78 | ||
| GenBank mRNA | BC064298.1 | - | 2 - 3 | 175 | 78 | |
| BC162974.1 | - | 2 - 3 | 235 | 78 | ||
| BC162996.1 | - | 2 - 3 | 235 | 78 | ||
| DQ278896.1 | - | 2 - 3 | 141 | 78 | ||
| EF426725.1 | - | 2 - 3 | 173 | 78 | ||
| EF426726.1 | - | 3 - 4 | 352 | 78 | ||
| EF426727.1 | - | 2 - 3 | 441 | 78 | ||
More Information
Set Membership: |
|

epoa
Genomic Map