Assay Details
| Gene Symbol: epoa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038009.2 | NP_001033098.1 | 4 - 5 | 695 | 78 | |
| NM_001115127.1 | NP_001108599.1 | 5 - 6 | 606 | 78 | ||
| NM_001115128.1 | NP_001108600.1 | 4 - 5 | 427 | 78 | ||
| GenBank mRNA | BC064298.1 | - | 4 - 5 | 429 | 78 | |
| BC162974.1 | - | 4 - 5 | 489 | 78 | ||
| BC162996.1 | - | 4 - 5 | 489 | 78 | ||
| DQ278896.1 | - | 4 - 5 | 395 | 78 | ||
| EF426725.1 | - | 4 - 5 | 427 | 78 | ||
| EF426726.1 | - | 5 - 6 | 606 | 78 | ||
| EF426727.1 | - | 4 - 5 | 695 | 78 | ||
More Information
Set Membership: |
|

epoa
Genomic Map