Assay Details
| Gene Symbol: abcg2d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042772.1 | NP_001036237.1 | 9 - 10 | 1305 | 105 | |
| GenBank mRNA | DQ223861.1 | - | 9 - 10 | 1305 | 105 | |
More Information
Set Membership: |
|

abcg2d
Genomic Map