Assay Details
| Gene Symbol: abcg2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042775.1 | NP_001036240.1 | 11 - 12 | 1475 | 72 | |
| XM_021469846.1 | XP_021325521.1 | 11 - 12 | 1512 | 72 | ||
| GenBank mRNA | DQ223858.1 | - | 11 - 12 | 1475 | 72 | |
More Information
Set Membership: |
|

abcg2a
Genomic Map