Assay Details
| Gene Symbol: jph1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044348.1 | NP_001037813.1 | 1 - 2 | 630 | 83 | |
| XM_005163433.4 | XP_005163490.1 | 1 - 2 | 658 | 83 | ||
| GenBank mRNA | BC117663.1 | - | 1 - 2 | 630 | 83 | |
More Information
Set Membership: |
|

jph1b
Genomic Map