Assay Details
| Gene Symbol: atp2b1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044757.1 | NP_001038222.1 | 5 - 6 | 1057 | 85 | |
| XM_009300256.3 | XP_009298531.1 | 5 - 6 | 1632 | 85 | ||
| XM_021474964.1 | XP_021330639.1 | 5 - 6 | 1138 | 85 | ||
| XM_021474965.1 | XP_021330640.1 | 5 - 6 | 1138 | 85 | ||
| XM_021474966.1 | XP_021330641.1 | 4 - 5 | 792 | 85 | ||
| GenBank mRNA | BC076309.1 | - | 5 - 6 | 85 | ||
| BC092356.1 | - | 5 - 6 | 1073 | 85 | ||
| BC115143.1 | - | 5 - 6 | 1081 | 85 | ||
| BC121780.1 | - | 5 - 6 | 85 | |||
| BC171622.1 | - | 4 - 5 | 894 | 85 | ||
| DQ242508.1 | - | 5 - 6 | 1055 | 85 | ||
| HM449162.1 | - | 4 - 5 | 771 | 85 | ||
More Information
Set Membership: |
|

atp2b1a
Genomic Map