Assay Details
| Gene Symbol: sash1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044819.1 | NP_001038284.1 | 17 - 18 | 2656 | 95 | |
| XM_021468431.1 | XP_021324106.1 | 17 - 18 | 4066 | 95 | ||
| XM_021468432.1 | XP_021324107.1 | 16 - 17 | 3965 | 95 | ||
| XM_021468433.1 | XP_021324108.1 | 17 - 18 | 3366 | 95 | ||
| XM_021468434.1 | XP_021324109.1 | 17 - 18 | 2817 | 95 | ||
| XM_021468435.1 | XP_021324110.1 | 17 - 18 | 3528 | 95 | ||
| XM_021468436.1 | XP_021324111.1 | 16 - 17 | 2723 | 95 | ||
| GenBank mRNA | BC171610.1 | - | 17 - 18 | 2541 | 95 | |
More Information
Set Membership: |
|

sash1a
Genomic Map