Assay Details
| Gene Symbol: fam102bb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044844.1 | NP_001038309.1 | 10 - 11 | 1363 | 76 | |
| XM_005160711.4 | XP_005160768.1 | 9 - 10 | 1335 | 76 | ||
| GenBank mRNA | BC171399.1 | - | 10 - 11 | 1218 | 76 | |
More Information
Set Membership: |
|

fam102bb
Genomic Map