Assay Details
| Gene Symbol: fam102bb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044844.1 | NP_001038309.1 | 2 - 3 | 554 | 64 | |
| XM_005160711.4 | XP_005160768.1 | 2 - 3 | 622 | 64 | ||
| XM_009294745.3 | XP_009293020.1 | 2 - 3 | 622 | 64 | ||
| GenBank mRNA | BC171399.1 | - | 2 - 3 | 409 | 64 | |
More Information
Set Membership: |
|

fam102bb
Genomic Map