Assay Details
| Gene Symbol: hcfc1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045064.1 | NP_001038529.1 | 20 - 21 | 4164 | 60 | |
| XM_005169304.4 | XP_005169361.1 | 20 - 21 | 4251 | 60 | ||
| XM_009306034.3 | XP_009304309.1 | 20 - 21 | 4231 | 60 | ||
| XM_009306035.3 | XP_009304310.1 | 20 - 21 | 4224 | 60 | ||
More Information
Set Membership: |
|

hcfc1a
Genomic Map