Assay Details
| Gene Symbol: hcfc1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045064.1 | NP_001038529.1 | 14 - 15 | 2448 | 85 | |
| XM_005169304.4 | XP_005169361.1 | 14 - 15 | 2466 | 85 | ||
| XM_009306034.3 | XP_009304309.1 | 14 - 15 | 2446 | 85 | ||
| XM_009306035.3 | XP_009304310.1 | 14 - 15 | 2439 | 85 | ||
| GenBank mRNA | BC139848.1 | - | 14 - 15 | 2390 | 85 | |
| BC142544.1 | - | 14 - 15 | 2390 | 85 | ||
More Information
Set Membership: |
|

hcfc1a
Genomic Map