Assay Details
| Gene Symbol: hcfc1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045064.1 | NP_001038529.1 | 15 - 16 | 2581 | 81 | |
| XM_005169304.4 | XP_005169361.1 | 15 - 16 | 2599 | 81 | ||
| XM_009306034.3 | XP_009304309.1 | 15 - 16 | 2579 | 81 | ||
| XM_009306035.3 | XP_009304310.1 | 15 - 16 | 2572 | 81 | ||
| GenBank mRNA | BC139848.1 | - | 15 - 16 | 2523 | 81 | |
| BC142544.1 | - | 15 - 16 | 2523 | 81 | ||
More Information
Set Membership: |
|

hcfc1a
Genomic Map