Assay Details
| Gene Symbol: hcfc1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045064.1 | NP_001038529.1 | 18 - 19 | 3774 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

hcfc1a| Gene Symbol: hcfc1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045064.1 | NP_001038529.1 | 18 - 19 | 3774 | 74 | |
Set Membership: |
|