Assay Details
| Gene Symbol: zdhhc14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045187.1 | NP_001038652.1 | 1 - 2 | 246 | 63 | |
| XM_005160351.4 | XP_005160408.1 | 1 - 2 | 1245 | 63 | ||
| XM_005160352.4 | XP_005160409.1 | 1 - 2 | 1245 | 63 | ||
More Information
Set Membership: |
|

zdhhc14
Genomic Map