Assay Details
| Gene Symbol: glg1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045395.1 | NP_001038860.1 | 6 - 7 | 1035 | 119 | |
| GenBank mRNA | BC122331.1 | - | 6 - 7 | 1035 | 119 | |
| BC161662.1 | - | 6 - 7 | 954 | 119 | ||
| BC164269.1 | - | 6 - 7 | 977 | 119 | ||
More Information
Set Membership: |
|

glg1b
Genomic Map