Assay Details
| Gene Symbol: glg1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045395.1 | NP_001038860.1 | 7 - 8 | 1215 | 86 | |
| GenBank mRNA | BC122331.1 | - | 7 - 8 | 1215 | 86 | |
| BC161662.1 | - | 7 - 8 | 1134 | 86 | ||
| BC164269.1 | - | 7 - 8 | 1157 | 86 | ||
More Information
Set Membership: |
|

glg1b
Genomic Map