Assay Details
| Gene Symbol: glg1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045395.1 | NP_001038860.1 | 9 - 10 | 1556 | 91 | |
| GenBank mRNA | BC122331.1 | - | 9 - 10 | 1556 | 91 | |
| BC161662.1 | - | 9 - 10 | 1475 | 91 | ||
| BC164269.1 | - | 9 - 10 | 1498 | 91 | ||
More Information
Set Membership: |
|

glg1b
Genomic Map