Assay Details
| Gene Symbol: ano10b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077382.1 | NP_001070850.1 | 8 - 9 | 1426 | 95 | |
| XM_021470507.1 | XP_021326182.1 | 9 - 10 | 1685 | 95 | ||
| XM_021470508.1 | XP_021326183.1 | 8 - 9 | 1545 | 95 | ||
| GenBank mRNA | BC124774.1 | - | 8 - 9 | 1426 | 95 | |
| BC165242.1 | - | 6 - 7 | 1356 | 95 | ||
More Information
Set Membership: |
|

ano10b
Genomic Map