Assay Details
| Gene Symbol: ano10b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077382.1 | NP_001070850.1 | 9 - 10 | 1508 | 69 | |
| XM_021470507.1 | XP_021326182.1 | 10 - 11 | 1767 | 69 | ||
| XM_021470508.1 | XP_021326183.1 | 9 - 10 | 1627 | 69 | ||
| GenBank mRNA | BC124774.1 | - | 9 - 10 | 1508 | 69 | |
| BC165242.1 | - | 7 - 8 | 1438 | 69 | ||
More Information
Set Membership: |
|

ano10b
Genomic Map