Assay Details
| Gene Symbol: scn4bb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077573.1 | NP_001071041.1 | 3 - 4 | 807 | 76 | |
| GenBank mRNA | BC146712.1 | - | 3 - 4 | 806 | 76 | |
| BC162278.1 | - | 3 - 4 | 687 | 76 | ||
| BC162294.1 | - | 3 - 4 | 687 | 76 | ||
| DQ471306.1 | - | 2 - 3 | 261 | 76 | ||
| DQ489732.1 | - | 3 - 4 | 807 | 76 | ||
More Information
Set Membership: |
|

scn4bb
Genomic Map