Assay Details
| Gene Symbol: ppm1la | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077600.1 | NP_001071068.1 | 3 - 4 | 1120 | 63 | |
| GenBank mRNA | BC125953.1 | - | 3 - 4 | 1120 | 63 | |
| BC164403.1 | - | 3 - 4 | 766 | 63 | ||
More Information
Set Membership: |
|

ppm1la
Genomic Map