Assay Details
| Gene Symbol: b4galnt1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080570.1 | NP_001074039.1 | 5 - 6 | 595 | 121 | |
| XM_005162256.4 | XP_005162313.1 | 5 - 6 | 696 | 121 | ||
| XM_021469816.1 | XP_021325491.1 | 5 - 6 | 689 | 121 | ||
| GenBank mRNA | BC129323.1 | - | 5 - 6 | 595 | 121 | |
More Information
Set Membership: |
|

b4galnt1a
Genomic Map