Assay Details
| Gene Symbol: sfr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082860.1 | NP_001076329.1 | 2 - 3 | 472 | 87 | |
| GenBank mRNA | BC133892.1 | - | 2 - 3 | 472 | 87 | |
More Information
Set Membership: |
|

sfr1
Genomic Map