Assay Details
| Gene Symbol: yrdc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083867.1 | NP_001077336.1 | 2 - 3 | 384 | 70 | |
| GenBank mRNA | BC134955.1 | - | 2 - 3 | 384 | 70 | |
More Information
Set Membership: |
|

yrdc
Genomic Map