Assay Details
| Gene Symbol: ccnl1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001089363.1 | NP_001082832.1 | 1 - 2 | 293 | 144 | |
| XM_005159222.4 | XP_005159279.1 | 1 - 2 | 329 | 144 | ||
| XM_005159223.4 | XP_005159280.1 | 1 - 2 | 329 | 144 | ||
| XM_005159224.2 | XP_005159281.1 | 1 - 2 | 194 | 144 | ||
| XM_009293814.1 | XP_009292089.1 | 1 - 2 | 330 | 144 | ||
| GenBank mRNA | BC057418.1 | - | 1 - 2 | 335 | 144 | |
| BC097120.1 | - | 1 - 2 | 297 | 144 | ||
| BC124101.1 | - | 1 - 2 | 292 | 144 | ||
| BC139645.1 | - | 1 - 2 | 293 | 144 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005159222.4, XM_005159223.4, XM_005159224.2, XM_009293814.1 transcripts.
Set Membership: |
|

ccnl1a
Genomic Map