Assay Details
| Gene Symbol: ccnl1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001089363.1 | NP_001082832.1 | 3 - 4 | 482 | 129 | |
| XM_005159224.2 | XP_005159281.1 | 3 - 4 | 383 | 129 | ||
| GenBank mRNA | BC097120.1 | - | 3 - 4 | 486 | 129 | |
| BC139645.1 | - | 3 - 4 | 482 | 129 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005159224.2 transcripts.
Set Membership: |
|

ccnl1a
Genomic Map