Assay Details
| Gene Symbol: ccnl1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001089363.1 | NP_001082832.1 | 5 - 6 | 666 | 106 | |
| XM_005159222.4 | XP_005159279.1 | 6 - 7 | 857 | 106 | ||
| GenBank mRNA | BC097120.1 | - | 5 - 6 | 670 | 106 | |
| BC124101.1 | - | 7 - 8 | 1018 | 106 | ||
| BC139645.1 | - | 5 - 6 | 666 | 106 | ||
More Information
Set Membership: |
|

ccnl1a
Genomic Map