Assay Details
| Gene Symbol: ccnl1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001089363.1 | NP_001082832.1 | 7 - 8 | 870 | 94 | |
| XM_005159222.4 | XP_005159279.1 | 8 - 9 | 1061 | 94 | ||
| XM_005159223.4 | XP_005159280.1 | 9 - 10 | 1075 | 94 | ||
| XM_005159224.2 | XP_005159281.1 | 8 - 9 | 785 | 94 | ||
| XM_009293814.1 | XP_009292089.1 | 8 - 9 | 2550 | 94 | ||
| GenBank mRNA | BC057418.1 | - | 8 - 9 | 2555 | 94 | |
| BC084688.1 | - | 2 - 3 | 288 | 94 | ||
| BC097120.1 | - | 7 - 8 | 874 | 94 | ||
| BC114231.1 | - | 9 - 10 | 1021 | 94 | ||
| BC124101.1 | - | 9 - 10 | 1222 | 94 | ||
| BC139645.1 | - | 7 - 8 | 870 | 94 | ||
More Information
Set Membership: |
|

ccnl1a
Genomic Map