Assay Details
| Gene Symbol: prdx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001089425.1 | NP_001082894.1 | 3 - 4 | 451 | 72 | |
| GenBank mRNA | BC134156.1 | - | 3 - 4 | 451 | 72 | |
More Information
Set Membership: |
|

prdx4
Genomic Map