Assay Details
| Gene Symbol: dap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098737.1 | NP_001092207.1 | 8 - 9 | 825 | 69 | |
| GenBank mRNA | BC142900.1 | - | 8 - 9 | 825 | 69 | |
More Information
Set Membership: |
|

dap3
Genomic Map