Assay Details
| Gene Symbol: stxbp5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099981.2 | NP_001093451.1 | 17 - 18 | 1919 | 58 | |
| XM_009294727.3 | XP_009293002.1 | 17 - 18 | 2338 | 58 | ||
| XM_009294728.3 | XP_009293003.1 | 17 - 18 | 2338 | 58 | ||
| XM_009294730.3 | XP_009293005.1 | 17 - 18 | 2338 | 58 | ||
| XM_021468429.1 | XP_021324104.1 | 17 - 18 | 2338 | 58 | ||
More Information
Set Membership: |
|

stxbp5a
Genomic Map