Assay Details
| Gene Symbol: stxbp5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099981.2 | NP_001093451.1 | 19 - 20 | 2132 | 79 | |
| XM_009294727.3 | XP_009293002.1 | 19 - 20 | 2551 | 79 | ||
More Information
Set Membership: |
|

stxbp5a
Genomic Map