Assay Details
| Gene Symbol: stxbp5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099981.2 | NP_001093451.1 | 25 - 26 | 3125 | 67 | |
| XM_009294727.3 | XP_009293002.1 | 25 - 26 | 3586 | 67 | ||
| XM_009294728.3 | XP_009293003.1 | 24 - 25 | 3538 | 67 | ||
| XM_009294730.3 | XP_009293005.1 | 23 - 24 | 3445 | 67 | ||
| XM_021468429.1 | XP_021324104.1 | 23 - 24 | 3487 | 67 | ||
More Information
Set Membership: |
|

stxbp5a
Genomic Map