Assay Details
| Gene Symbol: map7d2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001327937.1 | NP_001314866.1 | 2 - 3 | 890 | 112 | |
| XM_009301270.3 | XP_009299545.1 | 2 - 3 | 618 | 112 | ||
| XM_009301271.3 | XP_009299546.1 | 2 - 3 | 604 | 112 | ||
| XM_009301272.3 | XP_009299547.1 | 2 - 3 | 617 | 112 | ||
| XM_009301273.3 | XP_009299548.1 | 2 - 3 | 612 | 112 | ||
| XM_009301274.3 | XP_009299549.1 | 2 - 3 | 612 | 112 | ||
| XM_009301275.3 | XP_009299550.1 | 2 - 3 | 627 | 112 | ||
| XM_021475963.1 | XP_021331638.1 | 2 - 3 | 602 | 112 | ||
More Information
Set Membership: |
|

map7d2a
Genomic Map