Assay Details
| Gene Symbol: map7d2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001327937.1 | NP_001314866.1 | 3 - 4 | 984 | 85 | |
| XM_009301271.3 | XP_009299546.1 | 3 - 4 | 698 | 85 | ||
| XM_009301272.3 | XP_009299547.1 | 3 - 4 | 711 | 85 | ||
| XM_009301273.3 | XP_009299548.1 | 3 - 4 | 706 | 85 | ||
| XM_009301274.3 | XP_009299549.1 | 3 - 4 | 706 | 85 | ||
| XM_009301275.3 | XP_009299550.1 | 3 - 4 | 721 | 85 | ||
| XM_021475963.1 | XP_021331638.1 | 3 - 4 | 696 | 85 | ||
More Information
Set Membership: |
|

map7d2a
Genomic Map