Assay Details
| Gene Symbol: map7d2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009301273.3 | XP_009299548.1 | 6 - 7 | 1048 | 140 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

map7d2a| Gene Symbol: map7d2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009301273.3 | XP_009299548.1 | 6 - 7 | 1048 | 140 | |
Set Membership: |
|