Assay Details
| Gene Symbol: map7d2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001327937.1 | NP_001314866.1 | 9 - 10 | 1653 | 73 | |
| XM_009301270.3 | XP_009299545.1 | 9 - 10 | 1372 | 73 | ||
| XM_009301271.3 | XP_009299546.1 | 9 - 10 | 1367 | 73 | ||
| XM_009301272.3 | XP_009299547.1 | 8 - 9 | 1299 | 73 | ||
| XM_009301273.3 | XP_009299548.1 | 8 - 9 | 1252 | 73 | ||
| XM_009301274.3 | XP_009299549.1 | 7 - 8 | 1171 | 73 | ||
| XM_009301275.3 | XP_009299550.1 | 9 - 10 | 1390 | 73 | ||
| XM_021475963.1 | XP_021331638.1 | 9 - 10 | 1365 | 73 | ||
More Information
Set Membership: |
|

map7d2a
Genomic Map