Assay Details
| Gene Symbol: hbegfb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100401.2 | NP_001093871.2 | 2 - 3 | 376 | 95 | |
| XM_005161297.4 | XP_005161354.1 | 2 - 3 | 399 | 95 | ||
| GenBank mRNA | BC066705.1 | - | 2 - 3 | 306 | 95 | |
| BC150223.1 | - | 2 - 3 | 374 | 95 | ||
| EH542909.1 | - | 2 - 3 | 415 | 95 | ||
More Information
Set Membership: |
|

hbegfb
Genomic Map